Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Excellence Plus XP Microbalances

Hands-free operation and unmatched weighing performance. Mettler Toledo Excellence Plus XP Microbalances offer high speed, high performance and security features for measurements, users and data.

Manufacturer:  Mettler Toledo™ 11106048

 View more versions of this product

Catalog No. 01-910-670

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Includes: Automatic and detachable draft shield, automatic inner draft shield, ProFACT automatic calibration technology, RS-232-C interface, option slot for second interface (choice of 7 different interfaces), protective cover, detachable and adjustable terminal.

Description

Description

SmartGrid™ Weighing Pan

  • Minimizes effects of turbulence in the weighing chamber
  • Ensures more accurate results and shorter stabilization times
  • Smallest sample sizes can be weighed-in according to regulations
  • Increases the yield of expensive substances
SmartSens™ Sensors
  • For hands-free operation and personnel security Enable user to tare, open and close draft shield or print without touching the balance Promotes safe handling of samples, prevention of spills, minimized contamination risk
SmartScreen™ Display
  • Color-guided touchscreen for safe operation
  • Individual, color-coded profiles can be configured and documented for up to eight users or jobs
  • Protects users, data and processes, and guarantees seamless traceability
ErgoClips™
  • Facilitate secure placement and efficient handling of tare containers Farraday effect eliminates negative effects of statically charged tare containers
LevelControl and MinWeigh™
  • For full regulatory compliance LevelControl warns if balance is not correctly leveled MinWeigh signals when sample has not reached minimum weight defined by regulations

 

Standard Features

  • Prepared for weighing below the balance
  • User administration and password protection
  • Target weighing
  • Statistical evaluation
  • Formulation with security check
  • Density determination
  • Percent weighing
  • Piece counting with reference optimization
  • Alphanumeric data entry of four IDs

 

Warranty and Services

One year extendable to three years with return of warranty card.

Specifications

Specifications

Microbalance
0.001 mg
32.2cm
19.17 in.
3.5 sec.
26.3cm
19.17 x 10.35 x 12.67 in. (48.7 x 26.3 x 32.2cm)
GLP, ISO 9001
12.67 in.
Automatic and detachable draft shield, automatic i
48.7cm
10.35 in.
4cm
1.5 μg
Documents
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit