Learn More
Mettler Toledo™ Excellence Plus XPE Analytical Balances
Worry-free weighing thanks to balance intelligence
Manufacturer:  Mettler Toledo™ 30087700
 View more versions of this product
Catalog No. 01-911-002
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
XPE Analytical Balances provide the best performance in analytical weighing and support the highest requirements for safety, efficiency and ease of compliance. Thanks to low repeatability, XPE Analytical balances offer the smallest minimum weight. Quality Management features such as the innovative StatusLight™ and proprietary StaticDetect™ technology take the worry out of weighing and provide a high level of trust in your results. LevelControl and MinWeigh™ enable full regulatory compliance.
  • StatusLight™ uses color to help you identify the status of your balance, so you know exactly when it is ready for use
  • SmartGrid™ weighing pan minimizes effects of turbulence in weighing chamber and ensures more accurate results and shorter stabilization times
  • StaticDetect™ technology detects electrostatic charge on sample and/or its container; weighing error is measured and warning given if user-defined limit is exceeded
  • SmartSens sensors offer ergonomic, hands-free operation and personnel security enabling user to tare, open and close draft shield or print without touching balance
  • SmartScreen™ Display offers color-guided touchscreen for safe operation
  • Individual, color-coded profiles can be configured and documented for up to eight users or jobs, protecting users, data and processes, and guaranteeing seamless traceability
  • ErgoClips™facilitate secure placement and efficient handling of tare containers
  • LevelControl warns if balance is not correctly leveled
  • MinWeigh signals when a sample has not reached minimum weight defined by regulations
Specifications
Specifications
220g / 81g
0.8 A
3.07 in.
0.1 / 0.01 mg
50/60Hz
19.1 in.
1.5 sec.
100g
110/230V
11.3kg
7.2cm
0.25mg
1.5mg
12.75 in.
±0.15mg
<±0.06mg
26.3cm
Analytical balance
GLP, ISO 9001, UL, CSA
7.8cm
Color Graphic Display
Power supply, two free samples of the SmartPrep™
48.7cm
50 to 86°F
10 to 30°C
25 lb.
2.87 in.
110/230V 50/60Hz
48.7 x 26.3 x 32.5cm (19 x 10.3 x 12.75 in. )
RS-232C
32.5cm
XPE205DR
10.3 in.