Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Excellence Plus, XPE Series Micro-Analytical Balances Available on GSA/VA Contract for Federal Government customers only.

Designed to weigh small samples directly into large tare containers. Mettler Toledo Excellence Plus, XPE Series Micro-Analytical Balances are highly efficient models which allows for weighing down to one microgram while ensuring strict process tolerances.

Manufacturer:  Mettler Toledo™ 30105897

 View more versions of this product

Catalog No. 01-913-851


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

 
To ensure comprehensive weighing security, innovative features have been included in each micro-analytical balance model. All models offer quality management features. The innovative StatusLight™ uses color to indicate the status of the balance at a glance. The StaticDetect™ feature warns the user of potential weighing errors caused by static charge. This patented technology takes the worry out of weighing and provides users with a high level of trust in the results. The SmartGrid™ weighing pan, combined with the inner draft shield, minimizes the effects of air turbulence in the weighing chamber. Spilled samples are caught beneath the pan and do not affect the weighing results. Cross-contamination is reduced to a minimum. The LevelControl feature warns with an acoustic signal if the balance is not properly leveled, and provides corrective action in the display. The MinWeigh door allows dosing directly into the tare container through a small window, without opening the draft shield. To further reduce air turbulence in the weighing chamber, all micro-analytical balance models are equipped with an automatic dual draft shield. Outer doors open horizontally and inner doors open vertically, although both are individually configurable. Models 10147-674 and 10147-678 feature DeltaRange™ capabilities, which deliver ten times the readability anywhere within the balance range just by taring. 
 
All balances feature backlit color graphic display with touch screen operation. SmartSens programmable infrared sensors allow handsfree operation, reducing user error. To ensure the highest data security and full regulatory compliance, the SmartScreen touch screen offers individual profiles and access rights for up to eight users. The proFACT™ feature provides fully automatic time- and/or temperature-controlled internal adjustment and linearization. The operating menu can be viewed in 11 different languages and the operating software can be upgraded via the Internet. All models will display 12 different weighing units and available applications include statistics, formulation, piece counting, percent weighing, and density. Settings can be password protected. The level and the minimum weight per USP can be monitored by the user. A built-in RS-232 interface is standard, as well as a slot for a second interface (seven options available).
 
  • Capacities from 5.1 to 52g
  • Readabilities from 0.01 to 0.001mg
  • Features proFACT™ Fully Automatic Calibration Technology
  • Meets All GLP and ISO 9001 Requirements
  • Built-in RS-232 Interface

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Compliance

METTLER TOLEDO™ provides ISO 9001 manufacturing, nationwide ISO 9001 field service, ISO 9001 quality certification, and ISO 14001 environmental certification.

Specifications

Specifications

0.01mg
full range: 0.01 mg; fine range: 0.002 mg
100-240V AC, 0.8 A, 50/60 Hz, 12V DC 2.5 A
UL Listed. CE compliant.
2.5 / 3.5 sec.
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit