Learn More
Mettler Toledo™ XPE-L Series Precision Balances
Delivers best weighing performance for high capacity loads and provide a level of safety suitable for highly regulated environments
Manufacturer:  Mettler Toledo™ 30120456
 View more versions of this product
Catalog No. 01-913-876
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Durable and easy to clean
  • Offers high readability and repeatability for milligram to kilogram samples
  • High resolution technology with proFACT Advanced internal adjustment (on selected models)
  • SmartPan™ weighing pan with an integrated drip tray minimizes effects of air currents on weighing cell to deliver results up to two times faster
  • Repeatability values are improved up to two-fold
  • MonoblocHighSpeed weighing cell for guaranteed performance
  • IP54 in-use protection and a metal housing protects cell from dust, spills and damage from harsh chemicals
  • StatusLight™ indicates intuitively the status of the balance by using colors
  • LevelGuide™ provides warning when the balance is not level
  • Small footprint, ergonomic space-saver design
  • Large color touchscreen with big numbers on display
  • SmartTrac™ dosing guide
  • Compact draft shield has all-round access for handling large items
  • ErgoStand protects the display from spills and makes it easier to read
  • SmartSens infra-red sensors built into the terminal for touch-less operation
  • TestManager embedded software
  • MinWeigh function
  • FACT and GWP history
  • LabX™ software provides flexible SOP user guidance on the balance touchscreen
  • Calibry ready
  • RFID exchange with titrators
  • RFID SampleTrack
  • Built in RS232 with seven options for the second slot
Applications
Standard weighing, piece counting, percent weighing, statistics, formulation, dynamic weighing, density, differential weighing, factor calculation, RFID Exchange for titrators, RFID SampleTrack
Compliance
METTLER TOLEDO provides ISO 9001 manufacturing, nationwide ISO 9001 field service, ISO 9001 quality certification, and ISO 14001 environmental certification. UL listed. CSA certified.
Specifications
Specifications
10 kg
RS-232, Ethernet, Bluetooth, RFID
100 mg
80mg
Solid metal housing; stainless-steel weighing pan
100-240V AC, 0.8 A, 50/60 Hz, 12V DC 2.5 A
Precision Balance
UL Listed. CE compliant.
200 mg
XPE10001L
1 sec.