Learn More
Mettler Toledo™ Excellence, XPE Series Analytical Balances Available on GSA/VA Contract for Federal Government customers only.
Multicolor touch screen that guides the user for secure and ergonomic weighing of small sample sizes
Manufacturer:  Mettler Toledo™ 30131288
 View more versions of this product
Catalog No. 01-913-856
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
High-resolution analytical balances deliver all the statistics needed for Good Laboratory Practice, supporting the highest requirements for safety, efficiency, and ease of compliance. All models offer quality management features. The innovative StatusLight™ uses color to indicate the status of the balance at a glance. The StaticDetect™ feature warns the user of potential weighing errors caused by static charge. This patented technology takes the worry out of weighing and provides users with a high level of trust in the results. In addition, all models feature the unique SmartGrid™ weighing pan, which is supported at the rear of the weighing chamber, for more efficient weighing. In combination with the optional weighing kit, which includes ErgoClips™, the SmartGrid weighing pan becomes an ergonomic weighing system that can be precisely adapted to specific tasks and can help to increase productivity. 
The operating menu can be viewed in 11 different languages. The operating software can be upgraded via the Internet. All models will display 12 different weighing units and available applications include statistics, formulation, piece counting, percent weighing, and density. Settings can be password protected. The level and the minimum weight per USP can be monitored by the user. 
Ordering Information: The XPE Series Analytical balances are supplied with power supply, two free samples of the SmartPrep single-use antistatic weighing funnel, solid metal pan for placement atop the SmartGrid, a Static Detect drip tray, and ErgoClip™ basket.
  • Capacities from 120 to 520 g
  • Readabilities from 0.01 to 0.1 mg 
  • Meets all GLP and ISO 9001 Requirements
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Analytical Balance
0.1 mg
100-240V AC, 0.8 A, 50/60 Hz, 12V DC 2.5 A
XPE304
UL Listed. CE compliant.
1.5 sec.
0.4mg
0.08mg