Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Excellence XS Precision Balances

Designed for highest efficiency in everyday operation, reliabile and ease of use

Specifications

Material Die-cast Aluminum, Plastic, Chrome Steel
Electrical Requirements 100/240V, 50/60Hz
Display Type Color Graphic Display
Certifications/Compliance GLP, GMP, ISO 9001, ISO 14001, CE
View More Specs

Products 2
Catalog Number Mfr. No. Connection Type Capacity Readability Stabilization Time Price Quantity & Availability  
Catalog Number Mfr. No. Connection Type Capacity Readability Stabilization Time Price Quantity & Availability  
01-910-410
Accessories View Documents
Mettler Toledo™
11130183
23/s, bluetooth (wireless), ethernet, PS/2, USB 1.2 / 6.1 kg 0.01 / 0.1 g 1.2 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
01-910-407
Accessories View Documents
Mettler Toledo™
11130174
23/s, bluetooth (wireless), ethernet, PS/2, USB 4.1 kg 0.01 g 1.2 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

  • Fast, precise weighing results
  • Touch-screen display with graphical user guidance
  • MonoBloc High-Speed weighing cell, accelerated via dedicated processor
  • Alphanumeric sample ID entry (up to three IDs per sample)
  • GLP/GMP-compliant printouts
  • Verified versions and stand-alone platforms available
Design
  • Large, square weighing pans with up to 30% more surface area
  • Draft shield (1mg models) for direct access to weighing pan from any side
  • Robust construction and IP54 in-use protection


Applications:
Weighing, statistics, %-weighing, animal/dynamic weighing, piece counting, formulation and density determination

Warranty and Services

One year standard warranty with return of warranty card. Warranty can be extended for 12 additional months by ordering Cat. No. 09-313-190 or extended 24 additional months by ordering Cat. No. 01-910-600.

Specifications

Specifications

Die-cast Aluminum, Plastic, Chrome Steel
100/240V, 50/60Hz
Color Graphic Display
GLP, GMP, ISO 9001, ISO 14001, CE
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit