Learn More
Mettler Toledo™ Excellence XSE Analytical Balances Available on GSA/VA Contract for Federal Government customers only.
Worry-free weighing thanks to intuitive operation
Manufacturer:  Mettler Toledo™ 30087708
 View more versions of this product
Catalog No. 01-911-007
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
XSE Analytical Balances offer exceptional weighing performance and quality assurance features as well as excellent ergonomics. They are the perfect solution for a multitude of workplaces. Graphical LevelControl and MinWeigh™ enable full regulatory compliance. Innovative StatusLight™ feature takes the worry out of weighing. A green light lets you know when it's safe to start, indicating all tests are up-to-date and balance is level and functioning correctly.
  • StatusLight uses color to help you identify the status of your balance, so you know exactly when it is ready for use
  • SmartGrid™ weighing pan minimizes effects of turbulence in weighing chamber; ensures more accurate results and shorter stabilization times
  • SmartScreen™ display offers color-guided touchscreen for safe operation
  • Individual, color-coded profiles can be configured and documented for up to eight users or jobs, protecting users, data and processes, and guaranteeing seamless traceability
  • ErgoClips™ facilitate secure placement and efficient handling of tare containers
  • LevelControl warns if balance is not correctly leveled 
  • MinWeigh signals when sample has not reached minimum weight defined by regulations
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Analytical Balance
Touch screen
2.87 in.
0.1 mg
Power supply, two free samples of the SmartPrep&tr
45.3cm
50° to 86°F
10° to 30°C
25 lb.
3.07 in.
110/230V 50/60Hz
45.3 x 26.3 x 32.2cm
RS-232, RFID Ready
32.2cm
XSE104
10.37 in.
0.8 A
GLP, ISO 9001, UL, CSA
7.3cm
50/60Hz
17.83 in.
1.5 sec.
50g
110/230V
11.3kg
7.8cm
0.3mg
8mg
12.67 in.
±0.2mg
<±0.4mg
26.3cm