Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ FE30 FiveEasy™ Benchtop Conductivity Meters

Compact instrument for measurements in agriculture, food, beverage and water applications.

Manufacturer:  Mettler Toledo™ 51302912

 View more versions of this product

Catalog No. 01-915-663

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications Description & Specifications

Specifications

Package comes with meter, operating instructions, quick guide, power adapter
±0.5%
Autoranging
0 to 19.99psu
Automatic or Manual
0° to 100°C
±0.3°C
Mini-DIN
50/60Hz
ABS/PC Reinforced
100/240V 50/60Hz
0.1μS/cm to 199.9mS/cm
0.1mg/L to 199.9g/L
±5%
Conductivity
1
0.1°C
LCD
9VDC
100/240V
Conductivity, TDS, Salinity, Temperature
0.1μS/cm to 199.9mS/cm
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit