Mettler Toledo™ FG2 FiveGo™ Portable pH Meter

Compact instrument for pH measurement in agriculture, food, beverage and water applications.

Manufacturer: Mettler Toledo™ 51302906

 View more versions of this product

Catalog No. 01-917-157

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Display Type LCD
    IP Rating 54
    Weight (Metric) 0.18kg
    Height (English) 1.4 in.
    Accuracy (mV) ±1mV
    Calibration Points (pH) 1 or 2 Points, 3 pre defined buffer group
    Resolution (Temperature) 0.1°C
    Accuracy (Temperature) ±0.5°C
    Data Management Details 30 Measurements
    Inputs BNC for pH combination, Cinch/RCA jack for ATC probe
    Battery Type Four AAA
    Temperature (English) Operating 32° to 104°F
    Temperature (Metric) Operating 0 to 40°C
    Resolution (pH) 0.01
    Temperature (Metric) Range -0° to +100°C
    Range (mV) 0 to ±1999mV
    Resolution (mV) 1mV
    Probe Type NTC
    Includes Food Meter Kit: Meter, operating instructions, quick guide, electrode clip, wrist strap, four AAA batteries, LE427 puncture electrode (01-917-159), IP54 Connection covers, 2 each pH 4.01 and pH 7 Buffer sachets, 4 sample bottles, carry bag
    Weight (English) 0.4 lb.
    Range (pH) 0 to 14
    Height (Metric) 3.6cm
    Accuracy (pH) ±0.01
    Width (English) 3.2 in.
    Material ABS/PC Reinforced Window, Membrane Keypad
    Length (Metric) 16.9cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit