Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ FG3 FiveGo™ Portable Conductivity Meter

Compact instrument for conductivity measurement in agriculture, food, beverage and water applications.

Manufacturer:  Mettler Toledo™ 51302904

 View more versions of this product

Catalog No. 01-915-659

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications Description & Specifications

Specifications

Meter Kit
0.1 to 199.9μS/cm
0.1mg/L to 199.9g/L
0.1°C
LCD
Four AAA
180g
1.43 in.
0.1μS/cm to 199.9mS/cm
16.9cm
Conductivity
1.5V
Meter, operating instructions, quick guide, electrode clip, wrist strap, Four AAA batteries, LE703 conductivity electrode (01-915-679), IP54 connection covers, 2 each 1413μS/cm and 12.88mS/cm standard sachets
±0.5%
0 to 19.99psu
±0.3°C
Mini-DIN
54
1
3.6cm
Conductivity
ABS/PC Reinforced
0° to 50°C
3.25 in.
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit