FiveEasy Plus™ FEP20 pH Meter

Ideal for anyone who needs an economic and reliable solution for easy pH measurement

Manufacturer: Mettler Toledo 30089748

 View more versions of this product

Catalog No. 01-913-996

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Height (Metric) 52mm
    Width (Metric) 175mm
    Resolution (mV) 1mV
    Accuracy (mV) ±1mV
    Range (Temperature) 0° to 100°C
    Resolution (Temperature) 0.1°C
    Accuracy (Temperature) ±0.5°C
    Display Type LCD
    Data Management Details Up to 99 data storage
    Item Description FiveEasy Plus FEP20 pH Meter: TRIS Kit
    Outputs RS-232
    Includes Meter, operating manual, Quick guide, power adapter, LE420 pH electrode, electrode arm, 2X pH 4.01 and 2X pH 7.00 buffer sachets
    Certifications/Compliance CE marked, ISO 9001, ISO 14001
    Dimensions (L x W x H) 200 x 175 x 52mm
    Weight (Metric) 0.6kg
    Range (pH) 0.00 to 14.00
    Electrical Requirements 100 to 240V, 50 to 60Hz, 9VDC
    Resolution (pH) 0.01
    Length (Metric) 200mm
    Material ABS/PC
    Accuracy (pH) ±0.01
    Temperature Compensation Automatic or Manual
    Temperature Range (English) 32° to +212°F
    Temperature Range (Metric) 0° to +100°C
    Range (mV) -1999 to 1999mV

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit