Learn More
Mettler Toledo™ InLab™ Cables
Connects pH electrodes with Multipin to pH meters
Manufacturer:  Mettler Toledo™ 51344291
 View more versions of this product
Catalog No. 01-917-148
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
BNC/RCA (Cinch)
2m
Cable
78.7 in.
pH electrode