Learn More
Mettler Toledo™ InLab™ Reference Half Cells
For routine applications and measurements in ion-selective and pH half-cells. Mettler Toledo InLab InLab Reference Half Cells Protects against organic and corrosive samples and is ideal for optimum silver ion-free accuracy at various sample temperatures.
Manufacturer:  Mettler Toledo™ 51343192
 View more versions of this product
Catalog No. 01-911-496
Add to cart
Description
Description
  • Glass body
  • S7 screw head connector
  • Compatible with various cables for use with a wide range of meters
Specifications
Specifications
0.47 in.
3mol/L KCl
ARGENTHAL
Triple/Ceramic
4.72 in.
3mol/L KCl
0°C to 130°C
1.2 cm
For routine applications and measurements in Ion-selective half cells
S7
Glass
12 cm
32°F to 266°F
0°C to 130°C