Learn More
Mettler Toledo™ IPac Professional Installation and Qualification for Compact and KF Titrators
A fast and easy qualification solution.
Manufacturer:  Mettler Toledo™ B39990010V
 View more versions of this product
Catalog No. 01-911-607
Add to cart
Description
Description
Manufacturers of goods for end user consumption have to ensure that their products are safe and of consistent quality. Therefore qualification (IQ/OQ) of analytical and production equipment has long been a mandatory requirement in the pharmaceutical, biotechnology and medical industries, but today many customers in non-regulated environments also see the clear benefits of having their analytical equipment qualified. To support this customer group, Mettler Toledo has created the IPac for Compact and KF Titrators.
  • Professional installation and operational qualification (IQ/OQ) performed by fully authorized and factory-trained service personnel
  • Easy, fast and comprehensive through the qualification documentation handed out at the end of installation
  • IPac is regulatory-compliant instrument qualification
Proven performance for accurate results
  • Initial qualification consisting of IQ and OQ performed by a professional. General System Suitability Test completes the initial qualification by proving the reliability of the instrument and its proper functionality.
Productivity increase and cost saving
  • Professional installation and taking-to-operation make direct deployment and the immediate start of measurements possible.
Compliance
  • Documented qualification including calibration ensures compliance with quality systems such as GMP and ISO. IPac limits the risk of non-compliance during FDA audits and other quality management system assessments.
Tailor-made solutions
  • For every need, Mettler Toledo provides the right service solutions for life-long measurement performance, productivity and quality management.
Ordering Information: Please note pricing is for Zone 1; please contact your local representative for pricing in Zone 2 regions as additional travel fees may apply
Order Info
Please note pricing is for Zone 1; please contact your local representative for pricing in Zone 2 regions as additional travel fees may apply
Specifications
Specifications
Installation and qualification package
Professional installation and operational qualification (IQ/OQ) performed by fully authorized and factory-trained service personnel, easy, fast and comprehensive through the qualification documentation handed out at the end of installation, IPac is regulatory compliant instrument qualification
Zone 1 volumetric titrator