Learn More
Mettler Toledo™ LC-B LocalCAN Bidirectional Universal Data Interface
Manufacturer:  Mettler Toledo™ 228110
Catalog No. 01-911-153B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Lets AB (discontinued), PB (discontinued), SB, or College B (discontinued) Series balances interchange data with up to five peripheral devices at the same time with appropriate cables. Transfer data to a computer, use a foot switch, or activate a remote display—all simultaneously if needed. (See the following accessories: LC-P45 printer [01-911-152A]; LC-P43 printer [01-911-152B]; LC-PCalc printer [01-911-154D]; LocalCAN cables [01-911-153C through H, 01-913-77L]; Remote Control LocalCAN foot switch [01-911-153J]; and LocalCAN Auxiliary Remote Displays [01-910-123, 01-911-152D, 01-911-152E, 01-911-153A].)