Learn More
Mettler Toledo™ Excellence DM Digital Density Meters
Allows you to determine the density and specific gravity of a wide variety of liquids quickly and accurately. Mettler Toledo Excellence DM Digital Density Meters come with elegant ergonomic design, customizable user settings, and multilingual support making them more appealing and easy to use by a wide variety of operators.
Manufacturer:  Mettler Toledo™ 51337002
 View more versions of this product
Catalog No. 10376202
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.00004g/3
Touch Screen
30 sec.
0 to 91°C
Density meter
Model DM50
0 to 3g/3
1mL
Measure the density and specific gravity of a wide variety of liquids quickly and accurately
Promotions
Promotions
  • Mettler Toledo
    Purchase any instrument, and our data integrity experts will evaluate your data-handling process at no cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.
    06-30-2018
  • Mettler Toledo
    Purchase a METTLER TOLEDO™ balance or analytical instrument through Fisher Scientific™ and receive Triple Fisher Scientific Buck$ to use on your choice of any product(s) from Fisher Scientific.
    06-30-2017