Mettler Toledo™ Excellence RM Digital Benchtop Refractometers Available on GSA/VA Contract for Federal Government customers only.

Benchtop refractometers allow you to determine the refractive index, BRIX, HFCS, or user-defined concentrations of a wide variety of liquids quickly and accurately. Mettler Toledo Excellence RM Digital Benchtop Refractometer has an in-built Peltier temperature control which provides both reliable temperature stability and rapid heating/cooling rates.

Manufacturer: Mettler Toledo™ 51337004

 View more versions of this product

Catalog No. 10-376-204

  Check Availability
Add to Cart

    Specifications

    Description Model RM50
    Hertz 50/60Hz
    Measurement Time 20 sec.
    Refractive Index 1.32 to 1.58
    Accuracy 0.00002, 0.014 % Brix
    Scale Range 0 to 100% Brix
    Temperature Range (English) 41 to 167°F
    Temperature Range (Metric) 5 to 75°C

    • Inert sapphire prism facilitates measurement of thin, highly volatile, dark, or viscous liquids
    • Removable top section provides ease of cleaning with simple tap water without the need for tools
    • Plug-and-play connection of accessories such as the printer, barcode scanner, LogStraight™ biometric fingerprint reader, and ErgoSens™ infrared sensor for hands-free operation
    • Assisted sample loading and clearing are possible through the addition of FillPal™ and DryPal™ units
    • Optional EQPac (Equipment Qualification Package) provides comprehensive, detailed qualification guidance (IQ/OQ/PQ) to those working in regulated environments

    Expand Your Scope of Measurement
    • LiquiPhysics™ allows you to add parameters such as pH, color, and density/specific gravity to your refractive index readings
    • Simply connect a compatible SevenCompact™ pH or conductivity meter, colorimeter, or density cell to the RM module and define the method
    • Optional LabX™ LiquiPhysics™ Express software captures and manages all of these data points under a single platform
    • LabX™ also fully supports your CFR 21.11 requirements, provides user management, audit trail, flexible data export options, and fully customized reports
    Reliable Standards
    • Mettler Toledo Certified Combined Density and Refractive Index Standards reduce risk, uncertainty, and error-prone sample transfer Standards come complete with certificates including an easy-to-scan barcode containing all information necessary for calibration
    • Barcodes save you valuable time, reduce data entry error, and improve traceability

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit