Learn More
Mettler Toledo™ NewClassic MS Precision 0.01g Balances
Robust design, provides a visually attractive support for everyday weighing tasks
Specifications
Material Die-cast Aluminum, Stainless Steel, Plastic
Electrical Requirements 100/240V, 50/60Hz
Connection Type RS-232
Display Type LCD
Certifications/Compliance ISO 9001, ISO 14001, CE, UL, CSA
View More Specs
Products 2
Catalog Number Mfr. No. Model Capacity Readability Stabilization Time Price Quantity & Availability  
Catalog Number Mfr. No. Model Capacity Readability Stabilization Time Price Quantity & Availability  
01-920-30 View Documents Mettler Toledo™
11144950
MS802S 820 g 0.01 g 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
01-910-208 View Documents Mettler Toledo™
11144962
MS3002S 3.2 kg 0.01 g 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • HCD (High Contrast Display)
  • Metal High-grade chemical resistant die-cast housing
  • Fully automatic time and temperature-controlled internal adjustment (FACT)
  • Protective cover
  • Below weighing hook for weighing below the balance
  • MonoBloc™ weighing technology
  • Overload protection
  • Built in Date and Time function
  • SmartTrac shows the entire weighing range
  • Programmable 3 SmartKeys for shortcut access to applications
  • Protection of main balance configuration
Sensitivity temperature drift for models without FACT: 10 ppm/ °C. Stability of sensitivity as from first installation with FACT automatic adjustment switched on.
Specifications
Specifications
Die-cast Aluminum, Stainless Steel, Plastic
RS-232
ISO 9001, ISO 14001, CE, UL, CSA
100/240V, 50/60Hz
LCD