Learn More
Mettler Toledo™ NewClassic MS-S Draft Shield with Sliding Doors
Improves the quality of your weighing results, especially for balances with fine readability ranges (≤0.001g). Mettler Toledo NewClassic MS-S Draft Shield with Sliding Doors protects weighing samples from surrounding influences, resulting in optimized repeatability and fast stabilization times.
$844.00
Specifications
For Use With (Equipment) 0.1 mg and 1 mg MS and ML Balance
Type Draft Shield
View More Specs
Products 1
Catalog Number Mfr. No. Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Height (Metric) Price Quantity & Availability  
01-917-176
View Documents
Mettler Toledo™
12122404
23.7 cm
Each for $844.00
Add to cart
Description
Description
  • With left/right/top sliding doors
  • For 1mg/10mg top loading balances and 0.1mg/0.001g models
  • QuickLock feature allows all glass panels to be removed in just few simple steps for easy cleaning
Specifications
Specifications
0.1 mg and 1 mg MS and ML Balance
Draft Shield