Learn More
Mettler Toledo™ Bluetooth RS232 Serial Adapters for P50 Line Thermal Printers
For use with P50 Line Thermal Printers.
Manufacturer:  Mettler Toledo™ 30086495
 View more versions of this product
Catalog No. 01-910-274
Add to cart
Description
Description
Bluetooth RS232 Serial Adapter
  • Accessory for P-56RUE and P-58RUE Thermal printers
  • For wireless connection between printer and balance or between balance and PC
  • Single or paired
Specifications
Specifications
Bluetooth RS232 Serial Adapter
P-56RUE and P-58RUE Thermal printers
Pair