Learn More
Mettler Toledo™ Portable Dissolved Oxygen Meters
Includes: Ready-to-use field kit includes meter with mini-DIN connection, IP67-rated oxygen probe with 6 1/2 or 323/4 ft. long (2 or 10m) cable, sturdy plastic carrying case, batteries, instructions, instrument sealing kit, membrane caps, and membrane-cap fill solution.
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
01-920-21
|
Mettler Toledo™
MO1282M |
Cable length: 6.5 ft. | Cable length: 6.5 ft. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | View substitute products » |
01-920-22
|
Mettler Toledo™
MO12810M |
Cable length: 32.75 ft. | Cable length: 32.75 ft. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
For use in both lab and field environments—provide ruggedness and durability at an affordable price. Offer barometric compensation, salinity compensation, serial output and auto-endpoint and auto-off functions. So reliable, a battery change is the only internal maintenance ever needed. Ultrasonically welded casing is fully waterproofed to IP67 specifications—when used with special booted electrodes, the unit is dustproof and watertight to a depth of 3 1/4 ft. (1m) for 30 minutes. Battery life is 500 hours.
Order Info
Dimensions: 1 3/4L × 3 3/8W × 7 7/8 in.H (4.5 × 8.5 × 20cm). Net weight, 1 lb. (0.45kg).
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.