Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Routine Series pH and pH/ATC Combination Electrodes

Designed for accurate measurement in a wide range of sample types. With EQUITHAL? technology, Mettler Toledo Routine Series pH and pH/ATC Combination Electrodes guarantees shortest response times despite large changes in temperature.

Manufacturer:  Mettler Toledo™ 51343053

 View more versions of this product

Catalog No. 01-911-453

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications Description & Specifications

Specifications

0 to 100°C
0° to 100°C
Ceramic
170mm
S7 Screw head
ARGENTHAL
32° to 212°F
0.47 in.
3mol/L KCl
<20 sec.
0 to 14
Glass
3mol/L KCl
12mm
pH meter
6.69 in.
For accurate measurement in a wide range of sample types
pH combination electrode
HA Glass
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit