Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ S220 SevenCompact™ pH/Ion Benchtop Meter

Precise electrochemical measurement combined with flexibility, innovative design and ease-of-use. Mettler Toledo™ S220 SevenCompact pH/Ion Benchtop Meter measures ORP and ion concentration in various units and is suited for a wide range of applications. A single keypress is all that is required to start a measurement or calibration, toggle measurement modes or switch between displays.

Manufacturer:  Mettler Toledo™ 30019031

 View more versions of this product

Catalog No. 01-915-103


  Check Availability
Add to cart

Description

Description

Features
  • User friendly design: color display, intuitive icons and menu settings in 10 languages
  • uFocus™ view provides alternate display for essential reading/calculations
  • Pre-programmed settings for eight most commonly used ions
  • READ or CAL buttons initiate a measurement or calibration, with three options for endpointing
  • Intelligent Sensor Management (ISM™) for automatic recognition and secure, highly reproducible results
  • Professional calibration support includes pre- and user-defined buffer groups, calibration reminder with optional sensor blocking and a pH senor icon
  • GLP support functions: PIN protection, print-out and measurement limit monitoring
  • Acoustic signals for warnings, button presses, or end-point stability
  • Vertical uPlace™ movement for exacting electrode placement in sample
  • Storage for 1000 datasets; optional LabX direct software adds infinite data space
  • Comprehensive service package including IQ/OQ (sold separately)
  • Integrated USB and RS-232 interfaces for data exchange
  • Meter Kits (01-915-102, 103, 104) come with pH electrode/probe, InLab Pro-ISM
 
Temperature: an important parameter
  • The S220 supports both NTC30kW and Pt1000 temperature sensors, with option of automatic detection or manual selection 
  • Three options for temperature capture: integrated temperature sensor in electrode, external temperature sensor or setting temperature manually
  • Temperature measuring range: -30 to 130°C
Specifications

Specifications

pH/Ion
MTC: -30° to +130°C, ATC: -5° to +130°C
-2 to 20
±0.002
0.1/1mV
±0.1°C
9/12V
MTC: -86° to +266°F, ATC: -41° to +266°F
Meter, pH electrode/probe (InLab Routine Pro-ISM), pH guide, 2 buffer sachets for pH 4.01, 7, 9.21 and 10
0.001/0.01/0.1
2000 to 2000mV
±0.2mV
TFT
Meter Kit: S220-BIO
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit