Learn More
Mettler Toledo™ SevenGo™ perfectION Combined ISE Electrodes Available on GSA/VA Contract for Federal Government customers only.
Get an “All-In-One Solution” with SevenGo™ perfectION Combined ISE Electrodes. The brilliant Click & Clear™ press mechanism allows for quick cleaning and ensures stable readings even in difficult samples.
Manufacturer:  Mettler Toledo™ 51344803
 View more versions of this product
Catalog No. 01-928-252
Add to cart
Description
Description
  • As vast as the range of different ions, is the selection of ion­selective electrodes (ISE) 
  • The special Click & Clear™ junction allows for an optimal contact of the electrolyte solution and the sample
  • With the dedicated solutions the sample can be optimally prepared for successful measurement of ion concentration
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Ca+2 ions
ISA
32° to 104°F
Combined ISE Electrode
Lemo
0.02 to 40100mg/L
0° to 40°C