Mettler Toledo™ SG2 SevenGo™ pH Portable Meter

Built-in data memory and automatic functions for outstanding performance

Manufacturer: Mettler Toledo™ 51302523

 View all options for this product

Catalog No. 01-910-516

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Includes Wrist strap, four AA batteries, operating instructions, test certificate, declaration of conformity
    Range (pH) 0 to 14
    Resolution (pH) 0.01
    Accuracy (pH) ±0.01
    Range (mV) 0 to 1999mV
    Resolution (mV) 1mV
    Accuracy (mV) ±1mV
    Calibration Points (pH) 3
    Resolution (Temperature) 0.1°C
    Accuracy (Temperature) ±0.5°C
    Display Type LCD
    Data Points 30
    Inputs BNC
    Probe Type NTC
    Battery Type Four AA
    IP Rating 67
    Temperature (English) Operating 32° to 104°F
    Temperature (Metric) Operating 0 to 40°C
    Dimensions (L x W x H) 1.75 x 3.5 x 8.62 in. (45 x 90 x 220cm)
    Weight (English) 11.5 oz.
    Weight (Metric) 0.32kg
    For Use With (Application) pH
    Height (English) 8.62 in.
    Height (Metric) 220cm
    Length (English) 1.75 in.
    Width (English) 3.5 in.
    Width (Metric) 90cm
    Type pH Meter with Electrode, 1.2m Cable and Case
    Length (Metric) 45cm

    • Portable meter measures pH, mV and temperature
    • Designed for easy, comfortable right- or left-handed operation
    • Choice of three packaging configurations
    • Waterproof and dustproof to IP-67 standard
    • Large display of measured value and settings
    • Rotatable electrode clip mounts left or right for single-handed operation

    SevenGo SG2

    • IP-67 pH meter with 0 to 14 pH range for routine use
    • Continuous indication of electrode and battery status
    • Automatic endpoint and buffer recognition
    • Automatic temperature compensation
    • Data memory for 30 GLP data points
    • 3-point calibration with predefined or user-defined buffers

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit