Learn More
Mettler Toledo Temperature Calibration Kits
For use with halogen moisture analyzers. Mettler Toledo Temperature Calibration Kits are used to adjust halogen drying units to ensure consistent results.
$642.50
Specifications
For Use With (Equipment) | Halogen Moisture Analyzer |
---|---|
Type | Temperature Calibration Kit |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
01-909-385
![]() |
Mettler Toledo
214455 |
Adjustment disc with precision thermometer, sample |
Each for $642.50
|
|
Description
- Adjustment disc with precision thermometer
- Sample pan handler
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Halogen Moisture Analyzer | |
Temperature Calibration Kit |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.