Learn More
Mettler Toledo™ Replacement Parts for V20 Karl Fischer™ Autotitrator
For use with V20 Karl Fischer Autotitrators.
Manufacturer:  Mettler Toledo™ 51107668
 View more versions of this product
Catalog No. 01-910-732
Add to cart
Description
Description
  • Allows transporting sample to titrator conveniently, quickly and without losing material
  • Typical samples include chocolate melt/sauce, honey, mustard, cosmetics (creams), dairy products, technical greases
  • Ground glass joint exactly fits opening of the titration vessel
  • Before analysis, spoon is first tared, sample then weighed and Visco-Spoon inserted into opening of titrator, where it remains during entire titration
  • Sample's water content is completely extracted with solvent
  • Used with KF Titrators or KF titration stand DV705
Specifications
Specifications
Water content determination
V20 Karl Fischer titrators
Visco-Spoon™