Learn More
Mettler Toledo™ Weighing Scoops Available on GSA/VA Contract for Federal Government customers only.
For convenient transferring of powdered samples into narrow-necked flasks or bottles
Manufacturer:  Mettler Toledo™ 4508
 View more versions of this product
Catalog No. 01-914-38
Add to cart
Description
Description
Nickel alloy, scoop-shaped weighing dish.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
0.044 lb.
Weighing Boat
Ideal for convenient transferring of powdered samples into narrow-necked flasks or bottles
10.7cm
20g
0.044 lb., 20g
4.2 in.
Nickel Chromium Steel