Learn More
Mettler Toledo™ Weights for Balances Available on GSA/VA Contract for Federal Government customers only.
Weights for balances
Manufacturer:  Mettler Toledo™ 158391
 View more versions of this product
Catalog No. 01-915-140
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Weight with knob, One piece (monobloc)
OIML Class E2
1g
1g
Wooden box
1
With certificate, ISO17025, ISO9001, ISO14001 environmental standard
High Grade Stainless Steel
8kg/dm3 at 20°C
Cylindrical
Each