Learn More
Mettler Toledo™ Weights for Balances
Weights for balances
Manufacturer:  Mettler Toledo™ 158421
 View more versions of this product
Catalog No. 01-915-143
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Weight with knob, One piece (monobloc)
OIML Class E2
10g
10g
Wooden box
1
With certificate, ISO17025, ISO9001, ISO14001 environmental standard
Stainless Steel
8kg/dm3 at 20°C
Cylindrical
Each