Learn More
Mettler Toledo™ F1 Premium Cylindrical Knob Weights
Used in mass labs all over the world, and for calibration of analytical and micro balances
Manufacturer:  Mettler Toledo™ 158640
 View more versions of this product
Catalog No. 01-915-175
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Initial weight calibration uses robotic technology which excludes human error and leads to consistent and reproducible results with low uncertainty values Premium, vacuum melted steel offers an anticorrosive surface, and low magnetization and susceptibility values Monobloc design and electro polishing of surface provide maximum stability
Specifications
Specifications
Weight with knob, One piece (monobloc)
OIML Class F1
20g
20g
Wooden box
1
ISO17025, ISO9001, ISO14001 environmental standard
Stainless Steel
8kg/dm3 at 20°C
Cylindrical
Each