Mettler Toledo™ E2 Premium Wire Weights with Certificate Available on GSA/VA Contract for Federal Government customers only.

Individual weights

Manufacturer: Mettler Toledo™ 158331

 View more versions of this product

Catalog No. 01-915-134

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Quantity Each
    Product Type Wire weight
    Certifications/Compliance With certificate, ISO17025, ISO9001, ISO14001 environmental standard
    Includes Wooden box
    Classification OIML Class E2
    Packaging Quantity 1
    Material High Grade Stainless Steel
    Weight (Metric) 10mg
    Density 8kg/dm3 at 20°C
    Mass 10mg
    Shape Triangle

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit