Learn More
Mettler Toledo™ XP Series Precision Electronic Toploading Balances: Readability 100mg
Offers maximum security in regulated environments
Manufacturer:  Mettler Toledo™ 11130220
 View more versions of this product
Catalog No. 01-910-361
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Precision Balance
RS232
ISO 14001, GLP, CE
22.3cm
3.7 in.
50/60Hz
15.4 in.
±100mg
XP10001S
1 sec.
5 to 40°C
6.6kg
7.6 in.
19.4cm
39.2 x 19.4 x 9.6cm (15.4 x 7.6 x 3.7 in. )
120g
10,100g
0.8A
8.7 in.
Touch screen
100mg
9.6cm
AC adapter with country specific power cable, protective cover, operating instructions, production certificate, CE declaration of conformity
39.2cm
80mg
Die-cast Aluminum, Plastic, Stainless Steel
41 to 104°F
110/230V
14.6 lb.
7.4 in.
19cm
110/230V 50/60Hz
1 Year
Color Graphic Display
Accessories