Learn More
Mettler Toledo™ XS Precision L-Platform Balances
Offers the required high capacity and precision with large-sized platform
Manufacturer:  Mettler Toledo™ 11130669
 View more versions of this product
Catalog No. 01-910-461
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Chemical-resistant housing and robust design overload protection makes XS-L the ideal choice for everyday use in harsh environements.
  • MonoBloc™ HighSpeed weighing technology for fast and accurate results reduces settling time - stabilization time - up to 20%
  • FACT fully automatic temperature-controlled internal adjustment
  • Backlit graphic display with touchscreen operation
  • Weighing pan made from stainless steel, with draft ring or draft shield depending on resolution
  • Pacesetting interface (RS232) flexibility - including Ethernet, wireless Bluetooth and PS/2 - for efficient data capture and simple network integration
  • Feedthrough for below-the-balance weighing (optional)
  • Protective cover for terminal and weighing platform
  • Clear text dialogue in multiple languages
Specifications
Specifications
6.4 / 32.1 kg
RS-232, ethernet, wireless bluetooth, PS/2
CE, ISO 9001, ISO 14001
11 in.
1 g
13cm
FACT fully automatic
600/100mg
1.5 sec.
100/240V
27.3 lb.
11 in.
28cm
1 Year
Precision Balance
14 in.
28cm
35.56cm
5.1 in.
50/60Hz
±300mg
XS32001LDR
10kg
12.3kg
14.1 in.
35.8cm
100/240V, 50/60Hz
LCD
Documents
Accessories