Learn More
Mettler Toledo™ XS16000L Precision Balances L-Platform
Offers high capacitiy and precision with large-sized platform
Manufacturer:  Mettler Toledo™ 11130672
Catalog No. 01-910-462
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Chemical-resistant housing and robust design overload protection is ideal for everyday use in harsh environements.
  • MonoBloc™ HighSpeed weighing technology for fast and accurate results reduces settling time - stabilization time - up to 20%
  • FACT fully automatic temperature-controlled internal adjustment
  • Backlit graphic display with touchscreen operation
  • Weighing pan made from stainless steel, with draft ring or draft shield depending on resolution
  • Pacesetting interface (RS232) flexibility, including Ethernet, wireless Bluetooth™ and PS/2™, for efficient data capture and simple network integration
  • Feedthrough for below-the-balance weighing (option)
  • Protective cover for terminal and weighing platform
  • Clear text dialogue in multiple languages
Specifications
Specifications
16 kg
1 g
XS16000L
Documents
Accessories