Learn More
Medchemexpress LLC HY-17408 50mg Medchemexpress, Mevastatin CAS:73573-88-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1740850MG
Encompass_Preferred
Medchemexpress, HY-17408 50mg Mevastatin CAS:73573-88-3 Mevastatin (Compactin) is a first HMG-CoA reductase inhibitor that belongs to the statins class. Mevastatin is a lipid-lowering agent, and induces apoptosis, arrests cancer cells in G0/G1 phase. Mevastatin also increases endothelial nitric oxide synthase (eNOS) mRNA and protein levels. Mevastatin has antitumor activity and has the potential for cardiovascular diseases treatment[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0767