Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Mezlocillin sodium salt

Click to view available options
Quantity:
100mg
500mg
59798-30-0
C21H25N5NaO8S2
562.568
MFCD07793332
SGVORSZSYKDVFT-KZHFGBRHSA-N
123133909
(2R,5S,6S)-3,3-dimethyl-6-[[(2R)-2-[(3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)NC(=O)N4CCN(C4=O)S(=O)(=O)C)C(=O)O)C.[Na]
59798-30-0
C21H25N5NaO8S2
562.568
MFCD07793332
SGVORSZSYKDVFT-KZHFGBRHSA-N
123133909
(2R,5S,6S)-3,3-dimethyl-6-[[(2R)-2-[(3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)NC(=O)N4CCN(C4=O)S(=O)(=O)C)C(=O)O)C.[Na]

A broad spectrum antibiotic

Solubility Information Soluble in water or DMSO
Formula Weight 561.56
Physical Form Powder
Color White to Yellow
Quantity 500mg
Chemical Name or Material Mezlocillin sodium salt

storageNote1 : 2° to 8°C