Learn More
MilliporeSigma™ MF-Millipore™ Membrane Filter, 0.8 μm, Gridded Available on GSA/VA Contract for Federal Government customers only.
Mixed cellulose esters (MCE) membrane, hydrophilic
$135.90 - $291.50
Specifications
Autoclavable Not Autoclavable
Bubble Point ≥1.0 bar, air with water
Material Mixed Cellulose Esters (MCE)
Pore Size 0.8 μm
Sterility Non-sterile
View More Specs
Products 8
Catalog Number Mfr. No. Diameter (Metric) Color Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Color Price Quantity & Availability  
AABG01300
View Documents
MilliporeSigma™
AABG01300
13 mm Black
Pack of 100 for $181.30
Add to cart
AAWG02500
Accessories View Documents
MilliporeSigma™
AAWG02500
25 mm White
Pack of 100 for $152.80
Add to cart
AABG02500
Accessories View Documents
MilliporeSigma™
AABG02500
25 mm Black
Pack of 100 for $201.50
Add to cart
AAWG01300
View Documents
MilliporeSigma™
AAWG01300
13 mm White
Pack of 100 for $135.90
Add to cart
AABG04700 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
AABG04700
47 mm Black
Pack of 100 for $291.50
Add to cart
AAWG03700
View Documents
MilliporeSigma™
AAWG03700
37 mm White
Pack of 100 for $228.00
Add to cart
AABG03700
View Documents
MilliporeSigma™
AABG03700
Black
Pack for $239.00
Add to cart
AAWG04700 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
AAWG04700
47 mm White
Pack of 100 for $223.50
Add to cart
Description
Description
Mixed cellulose esters (MCE) membrane, hydrophilic
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.
MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features & Benefits:
  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
Specifications
Not Autoclavable
Mixed Cellulose Esters (MCE)
Non-sterile
190 mL/min x cm2
150 µm
Hydrophilic
≥1.0 bar, air with water
0.8 μm
Membrane
16 L/min/ cm2
Filter Membranes