Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFAP4 Antibody (12D11), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242638F
Description
MFAP4 Monoclonal specifically detects MFAP4 in Human samples. It is validated for ELISA.Specifications
MFAP4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
MFAP4 | |
Recombinant human MFAP4 (22-255aa) purified from E. coli | |
100 μL | |
Signal Transduction | |
4239 | |
Store at 4C in the dark. | |
IgG1 κ |
ELISA | |
12D11 | |
ELISA | |
microfibril-associated glycoprotein 4, microfibrillar-associated protein 4 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction