Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFG Tray Chemical-Resistant Multipurpose Trays and Pans
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
Supplier: MFG Tray 6011031537
Description
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
- Reinforced white thermoset polyester composite construction Resistant to most detergents, mild acid and alkaline solutions, corrosive salts and many reagents
- Withstand temperatures from -51° to +121°C (-60° to +250°F) continuous exposure, or 149°C (300°F) intermittent
- High load-bearing capacity
- Excellent dimensional stability Easily steam cleaned
Order Info
Available in 1 in. deep (2.5cm) trays, shallow pans from 1 1/4 to 2 3/4 in. deep (3 to 7cm), and ventilated stacking pans from 2 3/4 to 4 1/8 in. deep (7 to 10.5cm).
Specifications
26 x 26 x 1.25 in. (66 x 66 x 3 cm) | |
3 cm | |
Fiberglass | |
66 cm | |
26 in. | |
Square | |
-51°C to +121°C |
White | |
66 cm | |
Tray and Pan | |
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills | |
1.25 in. | |
-60°F to +250°F | |
26 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction