Learn More
MFG Tray Chemical-Resistant Multipurpose Trays and Pans
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
Manufacturer:  MFG Tray 6120031537
 View more versions of this product
Catalog No. 15-239-13
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1.5 in.
28 in.
Fiberglass
28 in.
White
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
-51° to 121°C
3.75cm
71cm
Multipurpose Tray and Shallow Pan
71cm
28 x 28 x 1.5 in. (71 x 71 x 3.75cm)
-60° to 250°F