Learn More
MFGE8 Mouse anti-Human, Clone: B656M, Abnova™
Mouse monoclonal antibody raised against human MFGE8.
Manufacturer:  Abnova Corporation MAB12684
Catalog No. 89-934-541
Add to cart
Description
Description
Specifications
Specifications
MFGE8
B656M
Mouse monoclonal antibody raised against human MFGE8.
In PBS (10mM sodium azide).
BA46/EDIL1/HsT19888/OAcGD3S/SED1/SPAG10/hP47
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunocytochemistry Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
MFGE8
HMFG Membrane tissue/cell preparation
Protein A purification
RUO
Primary
4240