Learn More
MFGE8 Mouse anti-Human, Clone: SPM291, Abnova™
Mouse monoclonal antibody raised against native human MFGE8.
Manufacturer:  Abnova Corporation MAB13108
Catalog No. 89-934-543
Add to cart
Description
Description
Specifications
Specifications
MFGE8
SPM291
Mouse monoclonal antibody raised against native human MFGE8.
In 10mM PBS.
BA46/EDIL1/HsT19888/OAcGD3S/SED1/SPAG10/hP47
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
MFGE8
Human milk fat globule membrane preparation.
Protein A/G purification
RUO
Primary
4240