Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFGE8 Mouse anti-Human, Clone: SPM291, Abnova™
Mouse monoclonal antibody raised against native human MFGE8.
Supplier: Abnova Corporation MAB13107
Specifications
MFGE8 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
BA46/EDIL1/HsT19888/OAcGD3S/SED1/SPAG10/hP47 | |
Mouse | |
Protein A/G purification | |
RUO | |
4240 | |
Store at 4°C. | |
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
SPM291 | |
Mouse monoclonal antibody raised against native human MFGE8. | |
In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
MFGE8 | |
Human milk fat globule membrane preparation. | |
100 μg | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction