Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MFI2 Recombinant Protein

Recombinant protein for gene encoding cell-surface glycoprotein found on melanoma cells

Manufacturer:  Abnova™ H00004241P0125

 View more versions of this product

Catalog No. 89-010-283


Add to cart

Description

Description

The protein shares sequence similarity and iron-binding properties with members of the transferrin superfamily. The importance of the iron binding function has not yet been identified. This gene resides in the same region of chromosome 3 as members of the transferrin superfamily. Alternative splicing results in two transcript variants.

  • Human MFI2 full-length ORF ( AAH01875, 21 a.a. - 302 a.a.) recombinant protein with GST-tag at N-terminal
  • Gene description: antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5
  • Theoretical molecular weight: 56.76kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification: glutathione sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced glutathione, pH: 8.0 in elution buffer

Sequence: MEVRWCATSDPEQHKCGNMSEAFREAGIQPSLLCVRGTSADHCVQLIAAQEADAITLDGGAIYEAGKEHGLKPVVGEVYDQEVGTSYYAVAVVRRSSHVTIDTLKGVKSCHTGINRTVGWNVPVGYLVESGRLSVMGCDVLKAVSDHFGGSCVPGAGETSYSESLCRLCRGDSPGEGVCDK SPLERYYDYSGAFRCLAEGAGDVAFVKHSTVLENTDESPSRRQTWTRSEEEEGECPAHEEARRTMRSSAGQAWKWAPVHRPQDESDKGEFGKRAKSRDMLG

Best use within three months from the date of receipt.

ELISA, western blotting (recombinant protein), antibody production, protein array

Specifications

Specifications

AAH01875
Solution
4241
MFI2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MFI2
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
56.76
8
Glutathione Sepharose 4 Fast Flow
25 ug
MEVRWCATSDPEQHKCGNMSEAFREAGIQPSLLCVRGTSADHCVQLIAAQEADAITLDGGAIYEAGKEHGLKPVVGEVYDQEVGTSYYAVAVVRRSSHVTIDTLKGVKSCHTGINRTVGWNVPVGYLVESGRLSVMGCDVLKAVSDHFGGSCVPGAGETSYSESLCRLCRGDSPGEGVCDKSPLERYYDYSGAFRCLAEGAGDVAFVKHSTVLENTDESPSRRQTWTRSEEEEGECPAHEEARRTMRSSAGQAWKWAPVHRPQDESDKGEFGKRAKSRDMLG
CD228/FLJ38863/MAP97/MGC4856/MTF1
MFI2
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit