Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFNG Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | MFNG |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179289
|
Novus Biologicals
NBP179289 |
100 μL |
Each of 1 for $436.00
|
|
Description
MFNG Polyclonal specifically detects MFNG in Human samples. It is validated for Western Blot.Specifications
MFNG | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
beta-1,3-N-acetylglucosaminyltransferase manic fringe, EC 2.4.1.222, manic fringe (Drosophila) homolog, manic fringe homolog (Drosophila), MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase, O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | |
MFNG | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
NP_002396 | |
4242 | |
Synthetic peptide directed towards the C terminal of human MFNG. Peptide sequence QLLRTAQLPEQVTLSYGVFEGKLNVIKLQGPFSPEEDPSRFRSLHCLLYP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title