Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MFSD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569623
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
MFSD1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
Specifications
MFSD1 | |
Polyclonal | |
Unconjugated | |
MFSD1 | |
1200003O06Rik; hypothetical protein LOC641486; major facilitator superfamily domain containing 1; major facilitator superfamily domain-containing protein 1; Major facilitator superfamily domain-containing protein 1-like protein; Mfsd1; mfsd1a; RGD1305751; SMAP4; SMAP-4; Smooth muscle cell-associated protein 4; UG0581B09; zgc:123187 | |
Rabbit | |
Affinity chromatography | |
RUO | |
64747 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H3U5 | |
MFSD1 | |
synthetic peptide directed towards the middle region of human MFSD1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction