Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFSD3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA563237

Catalog No. PIPA563237


Add to cart

Description

Description

Immunogen sequence: VFHLDTLGAS MDAGTILRGS ALL Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 61%.

The function of this protein has not been specifically defined.
Specifications

Specifications

MFSD3
Polyclonal
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96ES6
major facilitator superfamily domain-containing protein 3
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
113655
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Unconjugated
MFSD3
Liquid
MFSD3
Recombit protein corresponding to Human MFSD3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only