MFZ 10-7, Tocris™

Negative allosteric modulator at mGlu5

Manufacturer: R&D Systems 4438/50

Quantity 50mg

 View more versions of this product

Catalog No. 44-385-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C15H9FN2·HCl
    Formula Weight 272.7
    Synonym 3-Fluoro-5-[2-(6-methyl-2-pyridinyl)ethynyl]benzonitrile hydrochloride
    Chemical Name or Material MFZ 10-7

    Specifications

    Formula Weight 272.7
    Quantity 50mg
    CAS 1224431-15-5
    Synonym 3-Fluoro-5-[2-(6-methyl-2-pyridinyl)ethynyl]benzonitrile hydrochloride
    Recommended Storage Store at +4°C
    Assay Percent Range >99%
    Molecular Formula C15H9FN2·HCl
    Chemical Name or Material MFZ 10-7

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit