Learn More
MGAT2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 157721AP
Catalog No. 50-173-0466
Add to cart
Description
Description
The product of this gene is a Golgi enzyme catalyzing an essential step in the conversion of oligomannose to complex N-glycans. The enzyme has the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. Mutations in this gene may lead to carbohydrate-deficient glycoprotein syndrome, type II. The coding region of this gene is intronless. Transcript variants with a spliced 5 ft. UTR may exist, but their biological validity has not been determined.
Specifications
Specifications
MGAT2
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q10469, Q921V5
CDGS2, GlcNAc T II, GLCNACTII, GNT II, GNT2, MGAT2
Rabbit
IgG
150 μL
-20°C
Primary
217664, 4247
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MGAT2
Liquid
MGAT2
MGAT2 Fusion Protein Ag8423
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse